Imprimer

Anti-DDT Mouse Monoclonal Antibody [clone: OTI1A1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505603
ORIGTA505603EA 593 EUR
ORIGTA505603
Anti-DDT Mouse Monoclonal Antibody [clone: OTI1A1]
Anticorps
  • Recommended Dilutions: Western Blot: 1:200-8000; Immunochemistry: 1:150

Type: Primary
Antigen: DDT (D-dopachrome tautomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR