Imprimer

Anti-ROMO1 Mouse Monoclonal Antibody [clone: OTI5G8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505611
ORIGTA505611EA 616 EUR
ORIGTA505611
Anti-ROMO1 Mouse Monoclonal Antibody [clone: OTI5G8]
Anticorps
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ROMO1 (reactive oxygen species modulator 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR