Imprimer

Anti-MAP2K3 Mouse Monoclonal Antibody [clone: OTI6F8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505643EA 603 EUR
ORIGTA505643
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: OTI6F8]
Anticorps
  • Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR