Imprimer

Anti-RAB24 Mouse Monoclonal Antibody [clone: OTI4H10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505647
ORIGTA505647EA 616 EUR
ORIGTA505647
Anti-RAB24 Mouse Monoclonal Antibody [clone: OTI4H10]
Anticorps
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: RAB24 (RAB24, member RAS oncogene family)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR