Imprimer

Anti-RAB21 Mouse Monoclonal Antibody [clone: OTI6G1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505718
ORIGTA505718EA 593 EUR
ORIGTA505718
Anti-RAB21 Mouse Monoclonal Antibody [clone: OTI6G1]
Anticorps
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150

Type: Primary
Antigen: RAB21 (RAB21, member RAS oncogene family)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR