Imprimer

Anti-PIK3R5 Mouse Monoclonal Antibody [clone: OTI5B1]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505815
ORIGTA505815EA 621 EUR
ORIGTA505815
Anti-PIK3R5 Mouse Monoclonal Antibody [clone: OTI5B1]
Anticorps
Recommended Dilutions: Western Blot: 1:100-1000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3R5 (phosphoinositide-3-kinase, regulatory subunit 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR