Imprimer

Anti-MAPK13 Mouse Monoclonal Antibody [clone: OTI12B8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505855EA 603 EUR
ORIGTA505855
Anti-MAPK13 Mouse Monoclonal Antibody [clone: OTI12B8]
Anticorps
Recommended Dilutions: Western Blot: 1:1000-1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK13 (mitogen-activated protein kinase 13)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR