Imprimer

Anti-SYT4 Mouse Monoclonal Antibody [clone: OTI3B5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505864EA 581 EUR
ORIGTA505864
Anti-SYT4 Mouse Monoclonal Antibody [clone: OTI3B5]
Anticorps
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SYT4 (synaptotagmin IV)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR