Imprimer

Anti-RASSF8 Mouse Monoclonal Antibody [clone: OTI1F5]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505924
ORIGTA505924EA 616 EUR
ORIGTA505924
Anti-RASSF8 Mouse Monoclonal Antibody [clone: OTI1F5]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: RASSF8 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR