Imprimer

Anti-LZIC Mouse Monoclonal Antibody [clone: OTI5E3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505941EA 586 EUR
ORIGTA505941
Anti-LZIC Mouse Monoclonal Antibody [clone: OTI5E3]
Anticorps
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150

Type: Primary
Antigen: LZIC (leucine zipper and CTNNBIP1 domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR