Imprimer

Anti-CTAG1B Mouse Monoclonal Antibody [clone: OTI5G3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA505953EA 603 EUR
ORIGTA505953
Anti-CTAG1B Mouse Monoclonal Antibody [clone: OTI5G3]
Anticorps
Recommended Dilutions: Western Blot: 1:200-1:500

Type: Primary
Antigen: CTAG1B (cancer/testis antigen 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR