Imprimer

Anti-IL36G Mouse Monoclonal Antibody [clone: OTI6E4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA505995
ORIGTA505995EA 616 EUR
ORIGTA505995
Anti-IL36G Mouse Monoclonal Antibody [clone: OTI6E4]
Anticorps
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: IL36G (interleukin 36, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR