Imprimer

Anti-MAP2K1 Mouse Monoclonal Antibody [clone: OTI1A9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA506009EA 603 EUR
ORIGTA506009
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: OTI1A9]
Anticorps
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR