Imprimer

Anti-ERG Mouse Monoclonal Antibody [clone: OTI5D10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA506079EA 621 EUR
ORIGTA506079
Anti-ERG Mouse Monoclonal Antibody [clone: OTI5D10]
Anticorps
  • Recommended Dilutions: Western Blot: 1:200-4000; Immunofluorescence: 1:100

Type: Primary
Antigen: ERG (v-ets avian erythroblastosis virus E26 oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR