Imprimer

Anti-RARRES1 Mouse Monoclonal Antibody [clone: OTI1C9]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA506082EA 603 EUR
ORIGTA506082
Anti-RARRES1 Mouse Monoclonal Antibody [clone: OTI1C9]
Anticorps
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: RARRES1 (retinoic acid receptor responder (tazarotene induced) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR