Imprimer

Anti-ZSCAN21 Mouse Monoclonal Antibody [clone: OTI3D4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA506147EA 603 EUR
ORIGTA506147
Anti-ZSCAN21 Mouse Monoclonal Antibody [clone: OTI3D4]
Anticorps
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZSCAN21 (zinc finger and SCAN domain containing 21)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR