Imprimer

Anti-DNM1L Mouse Monoclonal Antibody [clone: OTI4A4]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ORIGTA506198EA 603 EUR
ORIGTA506198
Anti-DNM1L Mouse Monoclonal Antibody [clone: OTI4A4]
Anticorps
  • Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150

Type: Primary
Antigen: DNM1L (dynamin 1-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR