Imprimer

Anti-PTPRC Mouse Monoclonal Antibody [clone: OTI3C8]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA506566
ORIGTA506566EA 616 EUR
ORIGTA506566
Anti-PTPRC Mouse Monoclonal Antibody [clone: OTI3C8]
Anticorps
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR