Imprimer

Anti-POMC Mouse Monoclonal Antibody [clone: OTI1F3]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA506611
ORIGTA506611EA 616 EUR
ORIGTA506611
Anti-POMC Mouse Monoclonal Antibody [clone: OTI1F3]
Anticorps
Recommended Dilutions: Western Blot: 1:400-4000; Immunochemistry: 1:150

Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR