Imprimer

Anti-RAPGEF1 Mouse Monoclonal Antibody [clone: OTI1B2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA506822
ORIGTA506822EA 616 EUR
ORIGTA506822
Anti-RAPGEF1 Mouse Monoclonal Antibody [clone: OTI1B2]
Anticorps
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: RAPGEF1 (Rap guanine nucleotide exchange factor (GEF) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR