Imprimer

Anti-IL10RA Mouse Monoclonal Antibody [clone: OTI1G10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA506955
ORIGTA506955EA 616 EUR
ORIGTA506955
Anti-IL10RA Mouse Monoclonal Antibody [clone: OTI1G10]
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR