Imprimer

Anti-HPGD Mouse Monoclonal Antibody [clone: OTI1C12]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA600100
ORIGTA600100EA 616 EUR
ORIGTA600100
Anti-HPGD Mouse Monoclonal Antibody [clone: OTI1C12]
Anticorps
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: HPGD (hydroxyprostaglandin dehydrogenase 15-(NAD))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR