Imprimer

Anti-KIT Mouse Monoclonal Antibody [clone: OTI1E2]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA600223
ORIGTA600223EA 616 EUR
ORIGTA600223
Anti-KIT Mouse Monoclonal Antibody [clone: OTI1E2]
Anticorps
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: KIT (v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR