Imprimer

Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1B10]

Fournisseur: OriGene
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

TA600244
ORIGTA600244EA 616 EUR
ORIGTA600244
Anti-PECAM1 Mouse Monoclonal Antibody [clone: 1B10]
Anticorps
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR