Imprimer

Anti-HDAC4 Rabbit polyclonal antibody unconjugated

Fournisseur: Biorbyt
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87886-50EA 0 EUR
BIRBORB87886-50
Anti-HDAC4 Rabbit polyclonal antibody unconjugated
Anticorps
Immunochemistry: 1:50; Western Blot: 2 ?g/ml

Type: Primary
Antigen: HDAC4 (histone deacetylase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR