Imprimer

Anti-HMGN2 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87887-50EA 0 EUR
BIRBORB87887-50
Anti-HMGN2 Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:100-1:400; Immunochemistry: 1:50; Western Blot: 2 ?g/ml

Type: Primary
Antigen: HMGN2 (high mobility group nucleosomal binding domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR