Imprimer

Anti-ICOSLG Mouse Monoclonal Antibody

Fournisseur: Biorbyt
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87888-50EA 0 EUR
BIRBORB87888-50
Anti-ICOSLG Mouse Monoclonal Antibody
Anticorps
Immunochemistry: 1:50

Type: Primary
Antigen: ICOSLG (inducible T-cell co-stimulator ligand)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR