Imprimer

Anti-MCM7 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87895-50EA 0 EUR
BIRBORB87895-50
Anti-MCM7 Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 1:50

Type: Primary
Antigen: MCM7 (minichromosome maintenance complex component 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR