Imprimer

Anti-CCR10 Rabbit polyclonal antibody unconjugated

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87909-50EA 0 EUR
BIRBORB87909-50
Anti-CCR10 Rabbit polyclonal antibody unconjugated
Anticorps
Immunochemistry: 5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CCR10 (chemokine (C-C motif) receptor 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR