Imprimer

Anti-FPR1 Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB87913-50EA 0 EUR
BIRBORB87913-50
Anti-FPR1 Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 10 ?g/ml; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: FPR1 (formyl peptide receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR