Imprimer

Anti-SYNGR2 Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB88051-50EA 0 EUR
BIRBORB88051-50
Anti-SYNGR2 Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: SYNGR2 (synaptogyrin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR