Imprimer

Anti-OSTC Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BIRBORB88064-50EA 0 EUR
BIRBORB88064-50
Anti-OSTC Rabbit Polyclonal Antibody
Anticorps
Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: OSTC (oligosaccharyltransferase complex subunit (non-catalytic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR