Imprimer

Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11856-50EA 550 EUR
ABCAAB11856-50
Anti-IL10 Rat Monoclonal Antibody [clone: JES5-2A5]
Type: Primary
Antigen: IL10 (interleukin 10)
Clonality: monoclonal
Clone: JES5-2A5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR