Imprimer

Anti-C4A Rat Monoclonal Antibody [clone: 16D2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11863-50EA 719 EUR
ABCAAB11863-50
Anti-C4A Rat Monoclonal Antibody [clone: 16D2]
Anticorps
Type: Primary
Antigen: C4A (complement component 4A (Rodgers blood group))
Clonality: monoclonal
Clone: 16D2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR