Imprimer

Anti-C5AR1 Mouse Monoclonal Antibody [clone: S5/1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11867-50EA 729 EUR
ABCAAB11867-50
Anti-C5AR1 Mouse Monoclonal Antibody [clone: S5/1]
Anticorps
Type: Primary
Antigen: C5AR1 (complement component 5a receptor 1)
Clonality: monoclonal
Clone: S5/1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR