Imprimer

Anti-IL1RL2 Mouse Monoclonal Antibody [clone: 8.5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11868-50EA 698 EUR
ABCAAB11868-50
Anti-IL1RL2 Mouse Monoclonal Antibody [clone: 8.5]
Anticorps
Type: Primary
Antigen: IL1RL2 (interleukin 1 receptor-like 2)
Clonality: monoclonal
Clone: 8.5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR