Imprimer

Anti-Podoplanin / gp36 Hamster Monoclonal Antibody [clone: RTD4E10]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11936-100EA 948 EUR
ABCAAB11936-100
Anti-Podoplanin / gp36 Hamster Monoclonal Antibody [clone: RTD4E10]
Anticorps
Type: Primary
Antigen: Podoplanin / gp36
Clonality: monoclonal
Clone: RTD4E10
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR