Imprimer
Anti-E74 like factor 1 Rabbit Polyclonal Antibody
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB11956-50EA 550 EUR
ABCAAB11956-50
Anti-E74 like factor 1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: E74 like factor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human