Imprimer

Anti-NFIB / NF1B2 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11989-50EA 697 EUR
ABCAAB11989-50
Anti-NFIB / NF1B2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NFIB / NF1B2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR