Imprimer

Anti-SLC22A2 Mouse Monoclonal Antibody [clone: PT2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB11992-50EA 550 EUR
ABCAAB11992-50
Anti-SLC22A2 Mouse Monoclonal Antibody [clone: PT2]
Type: Primary
Antigen: SLC22A2 (solute carrier family 22 (organic cation transporter), member 2)
Clonality: monoclonal
Clone: PT2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR