Imprimer

Anti-RNA polymerase sigma 70 Mouse Monoclonal Antibody [clone: 2G10]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12088-50EA 596 EUR
ABCAAB12088-50
Anti-RNA polymerase sigma 70 Mouse Monoclonal Antibody [clone: 2G10]
Type: Primary
Antigen: RNA polymerase sigma 70
Clonality: monoclonal
Clone: 2G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: E. coli
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR