Imprimer

Anti-RNA polymerase II RPB11 Mouse Monoclonal Antibody [clone: 4Y11]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12090-50EA 565 EUR
ABCAAB12090-50
Anti-RNA polymerase II RPB11 Mouse Monoclonal Antibody [clone: 4Y11]
Type: Primary
Antigen: RNA polymerase II RPB11
Clonality: monoclonal
Clone: 4Y11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR