Imprimer

Anti-NFKBIE Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12136-50EA 550 EUR
ABCAAB12136-50
Anti-NFKBIE Rabbit Polyclonal Antibody
Type: Primary
Antigen: NFKBIE (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR