Imprimer

Anti-PTPIA2 / PTPRN Rabbit Polyclonal Antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12154-100EA 550 EUR
ABCAAB12154-100
Anti-PTPIA2 / PTPRN Rabbit Polyclonal Antibody
Type: Primary
Antigen: PTPIA2 / PTPRN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR