Imprimer

Anti-PRMT1 Mouse Monoclonal Antibody [clone: PRMT1-171]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12189-50EA 714 EUR
ABCAAB12189-50
Anti-PRMT1 Mouse Monoclonal Antibody [clone: PRMT1-171]
Anticorps
Type: Primary
Antigen: PRMT1 (protein arginine methyltransferase 1)
Clonality: monoclonal
Clone: PRMT1-171
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR