Imprimer

Anti-RBM39 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12326-50EA 682 EUR
ABCAAB12326-50
Anti-RBM39 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: RBM39 (RNA binding motif protein 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR