Imprimer

Anti-14-3-3 beta/zeta Mouse Monoclonal Antibody [clone: 22-IID8B]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12347-50EA 583 EUR
ABCAAB12347-50
Anti-14-3-3 beta/zeta Mouse Monoclonal Antibody [clone: 22-IID8B]
Type: Primary
Antigen: 14-3-3 beta/zeta (14-3-3 beta/zeta)
Clonality: monoclonal
Clone: 22-IID8B
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR