Imprimer

Anti-FGF16 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12471-50EA 550 EUR
ABCAAB12471-50
Anti-FGF16 Goat polyclonal antibody
Type: Primary
Antigen: FGF16 (fibroblast growth factor 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR