Imprimer

Anti-TPO Mouse Monoclonal Antibody [clone: MoAb47]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12500-500EA 698 EUR
ABCAAB12500-500
Anti-TPO Mouse Monoclonal Antibody [clone: MoAb47]
Anticorps
Type: Primary
Antigen: TPO (thyroid peroxidase)
Clonality: monoclonal
Clone: MoAb47
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR