Imprimer

Anti-G0 Protein alpha Mouse monoclonal antibody [clone: 1C.2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB12522-25EA 550 EUR
ABCAAB12522-25
Anti-G0 Protein alpha Mouse monoclonal antibody [clone: 1C.2]
Type: Primary
Antigen: G0 Protein alpha
Clonality: monoclonal
Clone: 1C.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR